Assay Details
| Gene Symbol: ZKSCAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288592.1 | NP_001275521.1 | 5 - 6 | 1441 | 91 | |
| GenBank mRNA | BC148260.1 | - | 5 - 6 | 1172 | 91 | |
More Information
Set Membership: |
|

ZKSCAN7
Genomic Map