Assay Details
| Gene Symbol: ANAPC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002245.2 | NP_001002245.1 | 2 - 3 | 243 | 80 | |
| NM_001002246.2 | NP_001002246.1 | 3 - 4 | 306 | 80 | ||
| NM_001002247.2 | NP_001002247.1 | 2 - 3 | 251 | 80 | ||
| NM_001002248.2 | NP_001002248.1 | 3 - 4 | 314 | 80 | ||
| NM_001002249.2 | NP_001002249.1 | 3 - 4 | 365 | 80 | ||
| NM_001289414.1 | NP_001276343.1 | 3 - 4 | 247 | 80 | ||
| NM_001289415.1 | NP_001276344.1 | 2 - 3 | 273 | 80 | ||
| NM_001289416.1 | NP_001276345.1 | 2 - 3 | 302 | 80 | ||
| NM_001289417.1 | NP_001276346.1 | 4 - 5 | 398 | 80 | ||
| NM_016476.11 | NP_057560.8 | 3 - 4 | 336 | 80 | ||
| GenBank mRNA | AF151048.1 | - | 3 - 4 | 281 | 80 | |
| AF247565.1 | - | 2 - 3 | 181 | 80 | ||
| AF247789.1 | - | 3 - 4 | 259 | 80 | ||
| AK307214.1 | - | 3 - 4 | 306 | 80 | ||
| AK308091.1 | - | 1 - 2 | 157 | 80 | ||
| AW965775.1 | - | 3 - 4 | 247 | 80 | ||
| BC066308.1 | - | 3 - 4 | 350 | 80 | ||
| BC095454.1 | - | 3 - 4 | 265 | 80 | ||
| BC104641.1 | - | 2 - 3 | 199 | 80 | ||
| BC171892.1 | - | 3 - 4 | 248 | 80 | ||
| BC171898.1 | - | 2 - 3 | 215 | 80 | ||
| BC171899.1 | - | 3 - 4 | 256 | 80 | ||
| BC171900.1 | - | 2 - 3 | 185 | 80 | ||
| BG707824.1 | - | 3 - 4 | 297 | 80 | ||
| BU554167.1 | - | 3 - 4 | 316 | 80 | ||
| BU556841.1 | - | 3 - 4 | 280 | 80 | ||
| BY796995.2 | - | 2 - 3 | 166 | 80 | ||
| CA335305.1 | - | 4 - 5 | 361 | 80 | ||
| HY067392.1 | - | 2 - 3 | 244 | 80 | ||
More Information
Set Membership: |
|

ANAPC11
Genomic Map