Assay Details
| Gene Symbol: MISP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291291.1 | NP_001278220.1 | 1 - 2 | 867 | 102 | |
| GenBank mRNA | BC011002.2 | - | 1 - 2 | 867 | 102 | |
More Information
Set Membership: |
|

MISP3
Genomic Map