Assay Details
| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291822.1 | NP_001278751.1 | 1 - 2 | 150 | 130 | |
| NM_001291823.1 | NP_001278752.1 | 1 - 2 | 150 | 130 | ||
| NM_016523.2 | NP_057607.1 | 1 - 2 | 150 | 130 | ||
| NR_120305.1 | 1 - 2 | 150 | 130 | |||
| XM_017019414.1 | XP_016874903.1 | 1 - 2 | 269 | 130 | ||
| XM_017019415.1 | XP_016874904.1 | 1 - 2 | 328 | 130 | ||
| GenBank mRNA | AF175206.1 | - | 1 - 2 | 150 | 130 | |
| AF175207.1 | - | 1 - 2 | 150 | 130 | ||
| AF267244.1 | - | 1 - 2 | 150 | 130 | ||
| AF267245.1 | - | 1 - 2 | 150 | 130 | ||
| AJ305370.1 | - | 1 - 2 | 133 | 130 | ||
| BC096734.1 | - | 1 - 2 | 110 | 130 | ||
| BC098166.1 | - | 1 - 2 | 110 | 40015535 | 130 | |
| BC098257.1 | - | 1 - 2 | 110 | 40015534 | 130 | |
| BC098354.1 | - | 1 - 2 | 110 | 40015531 | 130 | |
More Information
Set Membership: |
|

KLRF1
Genomic Map