Assay Details
| Gene Symbol: LOC100129940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292024.1 | NP_001278953.1 | 2 - 3 | 598 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100129940| Gene Symbol: LOC100129940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292024.1 | NP_001278953.1 | 2 - 3 | 598 | 69 | |
Set Membership: |
|