Assay Details
| Gene Symbol: MAPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292040.1 | NP_001278969.1 | 2 - 3 | 1548 | 82 | |
| NM_002747.3 | NP_002738.2 | 2 - 3 | 1548 | 82 | ||
| XM_005258299.3 | XP_005258356.1 | 2 - 3 | 5910 | 82 | ||
| XM_011526074.2 | XP_011524376.1 | 2 - 3 | 6092 | 82 | ||
| XM_011526075.2 | XP_011524377.1 | 2 - 3 | 1550 | 82 | ||
| XM_011526076.2 | XP_011524378.1 | 2 - 3 | 1913 | 82 | ||
| XM_017025839.1 | XP_016881328.1 | 2 - 3 | 1481 | 82 | ||
| GenBank mRNA | AK295058.1 | - | 2 - 3 | 1469 | 82 | |
| AK303601.1 | - | 1 - 2 | 210 | 82 | ||
| BC040493.1 | - | 2 - 3 | 1520 | 82 | ||
| BC050299.1 | - | 2 - 3 | 1548 | 5303135 | 82 | |
| BC117216.1 | - | 1 - 2 | 602 | 40125767 | 82 | |
| BC143769.1 | - | 1 - 2 | 600 | 82 | ||
| X59727.1 | - | 1 - 2 | 1085 | 82 | ||
More Information
Set Membership: |
|

MAPK4
Genomic Map