Assay Details
| Gene Symbol: MAPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292040.1 | NP_001278969.1 | 3 - 4 | 1693 | 118 | |
| GenBank mRNA | BC040493.1 | - | 3 - 4 | 1665 | 118 | |
| BC143769.1 | - | 2 - 3 | 745 | 118 | ||
More Information
Set Membership: |
|

MAPK4
Genomic Map