Assay Details
| Gene Symbol: TRIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302694.1 | NP_001289623.1 | 1 - 2 | 254 | 69 | |
| XM_006714159.3 | XP_006714222.1 | 1 - 2 | 963 | 69 | ||
| XM_017007946.1 | XP_016863435.1 | 1 - 2 | 960 | 69 | ||
| GenBank mRNA | BP310511.1 | - | 1 - 2 | 253 | 69 | |
| DA310963.1 | - | 1 - 2 | 239 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302694.1, XM_006714159.3, XM_017007946.1 transcripts.
Set Membership: |
|

TRIM2
Genomic Map