Assay Details
Gene Symbol: C6orf99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302839.1 | NP_001289768.1 | 1 - 2 | 443 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302839.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
