Assay Details
Gene Symbol: C6orf99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302840.1 | NP_001289769.1 | 2 - 3 | 203 | 74 | |
GenBank mRNA | HY052980.1 | - | 2 - 3 | 204 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302840.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
