Assay Details
| Gene Symbol: UCMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303119.1 | NP_001290048.1 | 1 - 2 | 127 | 74 | |
| GenBank mRNA | JX169865.1 | - | 1 - 2 | 86 | 74 | |
More Information
Set Membership: |
|

UCMA
Genomic Map