Assay Details
| Gene Symbol: OVOL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303461.1 | NP_001290390.1 | 3 - 4 | 714 | 87 | |
| NM_001303462.1 | NP_001290391.1 | 2 - 3 | 303 | 87 | ||
| NM_021220.3 | NP_067043.2 | 3 - 4 | 763 | 87 | ||
| GenBank mRNA | AK022284.1 | - | 3 - 4 | 758 | 87 | |
| AL079276.1 | - | 2 - 3 | 343 | 87 | ||
| BC006148.1 | - | 3 - 4 | 763 | 3959436 | 87 | |
| BE791524.1 | - | 3 - 4 | 576 | 87 | ||
| BM563561.1 | - | 2 - 3 | 303 | 87 | ||
| BT007295.1 | - | 3 - 4 | 515 | 87 | ||
More Information
Set Membership: |
|

OVOL2
Genomic Map