Assay Details
| Gene Symbol: CENPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002876.2 | NP_001002876.1 | 4 - 5 | 398 | 117 | |
| NM_001304373.1 | NP_001291302.1 | 3 - 4 | 511 | 117 | ||
| GenBank mRNA | BE789455.1 | - | 3 - 4 | 443 | 117 | |
| BE797487.1 | - | 4 - 5 | 352 | 117 | ||
| BX332140.2 | - | 3 - 4 | 449 | 116 | ||
| BX417558.2 | - | 3 - 4 | 450 | 116 | ||
More Information
Set Membership: |
|

CENPM
Genomic Map