Assay Details
| Gene Symbol: MED11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305000.1 | NP_001291929.1 | 2 - 3 | 293 | 97 | |
| GenBank mRNA | BG503911.1 | - | 2 - 3 | 274 | 97 | |
More Information
Set Membership: |
|

MED11
Genomic Map