Assay Details
| Gene Symbol: TMPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032283.2 | NP_001027454.1 | 1 - 2 | 578 | 95 | |
| NM_001032284.2 | NP_001027455.1 | 1 - 2 | 578 | 95 | ||
| NM_001307975.1 | NP_001294904.1 | 1 - 2 | 578 | 95 | ||
| NM_003276.2 | NP_003267.1 | 1 - 2 | 578 | 95 | ||
| XM_005269132.3 | XP_005269189.1 | 1 - 2 | 569 | 95 | ||
| GenBank mRNA | AB209297.1 | - | 1 - 2 | 515 | 95 | |
| AF070631.1 | - | 1 - 2 | 519 | 95 | ||
| AK315221.1 | - | 1 - 2 | 507 | 96 | ||
| BC053675.1 | - | 1 - 2 | 524 | 6170639 | 95 | |
| EF028063.1 | - | 1 - 2 | 299 | 95 | ||
| S76736.1 | - | 1 - 2 | 483 | 95 | ||
| U09086.1 | - | 1 - 2 | 487 | 95 | ||
| U09087.1 | - | 1 - 2 | 520 | 95 | ||
| U09088.1 | - | 1 - 2 | 520 | 95 | ||
More Information
Set Membership: |
|

TMPO
Genomic Map