Assay Details
| Gene Symbol: HIST1H2BK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001312653.1 | NP_001299582.1 | 1 | 416 | 88 | |
| GenBank mRNA | AK292062.1 | - | 1 | 413 | 88 | |
More Information
Set Membership: |
|

HIST1H2BK
Genomic Map