Assay Details
| Gene Symbol: HMGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313892.1 | NP_001300821.1 | 1 - 2 | 163 | 99 | |
| GenBank mRNA | AV763680.1 | - | 1 - 2 | 163 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001313892.1 transcripts.
Set Membership: |
|

HMGB1
Genomic Map