Assay Details
| Gene Symbol: HMGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313893.1 | NP_001300822.1 | 1 - 2 | 923 | 84 | |
| GenBank mRNA | AK122825.1 | - | 1 - 2 | 925 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK122825.1, NM_001313893.1 transcripts.
Set Membership: |
|

HMGB1
Genomic Map