Assay Details
| Gene Symbol: RPL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315531.1 | NP_001302460.1 | 1 - 2 | 301 | 76 | |
| GenBank mRNA | BI596427.1 | - | 1 - 2 | 306 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001315531.1 transcripts.
Set Membership: |
|

RPL26
Genomic Map