Assay Details
| Gene Symbol: ALG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015050.2 | NP_001015050.2 | 1 - 2 | 151 | 61 | |
| XM_017005847.1 | XP_016861336.1 | 1 - 2 | 41 | 61 | ||
| GenBank mRNA | BC073816.1 | - | 1 - 2 | 146 | 5419254 | 61 |
| CD685738.1 | - | 1 - 2 | 172 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC073816.1, NM_001015050.2, XM_017005847.1 transcripts.
Set Membership: |
|

ALG1L
Genomic Map