Assay Details
| Gene Symbol: TAF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015892.1 | NP_001015892.1 | 2 - 3 | 456 | 119 | |
| NM_003187.4 | NP_003178.1 | 2 - 3 | 219 | 119 | ||
| GenBank mRNA | AI879232.1 | - | 2 - 3 | 176 | 119 | |
| BC003400.1 | - | 2 - 3 | 147 | 3448836 | 119 | |
| BC033320.1 | - | 2 - 3 | 1007 | 4837792 | 119 | |
| BG490028.1 | - | 2 - 3 | 279 | 119 | ||
| BG720149.1 | - | 2 - 3 | 199 | 119 | ||
| CX784292.1 | - | 2 - 3 | 258 | 119 | ||
| U21858.1 | - | 2 - 3 | 136 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC003400.1, BC033320.1, NM_001015892.1, NM_003187.4, U21858.1 transcripts.
Set Membership: |
|

TAF9
Genomic Map