Assay Details
| Gene Symbol: RPS15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018.4 | NP_001009.1 | 1 - 2 | 62 | 90 | |
| GenBank mRNA | BU602036.1 | - | 1 - 2 | 63 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001018.4 transcripts.
Set Membership: |
|

RPS15
Genomic Map