Assay Details
| Gene Symbol: PLAC8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029869.1 | NP_001025040.1 | 3 - 4 | 456 | 75 | |
| XM_005268381.3 | XP_005268438.1 | 3 - 4 | 861 | 75 | ||
| XM_011537571.2 | XP_011535873.1 | 4 - 5 | 949 | 75 | ||
| XM_017009137.1 | XP_016864626.1 | 3 - 4 | 525 | 75 | ||
| GenBank mRNA | BC130586.1 | - | 3 - 4 | 442 | 40146617 | 75 |
| BC130588.1 | - | 3 - 4 | 442 | 40146619 | 75 | |
| BC144387.1 | - | 3 - 4 | 407 | 75 | ||
| BC144389.1 | - | 4 - 5 | 505 | 75 | ||
| BC144392.1 | - | 3 - 4 | 419 | 75 | ||
More Information
Set Membership: |
|

PLAC8L1
Genomic Map