Assay Details
| Gene Symbol: SNRPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317169.1 | NP_001304098.1 | 3 - 4 | 218 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001317169.1 transcripts.
Set Membership: |
|

SNRPG
Genomic Map