Assay Details
| Gene Symbol: SNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317192.1 | NP_001304121.1 | 4 - 5 | 456 | 78 | |
| NM_001317193.1 | NP_001304122.1 | 3 - 4 | 405 | 78 | ||
| NM_007241.3 | NP_009172.2 | 4 - 5 | 456 | 78 | ||
| NR_133679.1 | 4 - 5 | 456 | 78 | |||
| GenBank mRNA | AF156102.1 | - | 4 - 5 | 405 | 78 | |
| BC008976.1 | - | 4 - 5 | 406 | 3452153 | 78 | |
| BC038830.1 | - | 4 - 5 | 430 | 3900806 | 78 | |
| BE818533.1 | - | 3 - 4 | 259 | 78 | ||
| BG248541.1 | - | 4 - 5 | 445 | 78 | ||
| BP342490.1 | - | 4 - 5 | 456 | 78 | ||
More Information
Set Membership: |
|

SNF8
Genomic Map