Assay Details
| Gene Symbol: RPL26L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317982.1 | NP_001304911.1 | 1 - 2 | 57 | 70 | |
| GenBank mRNA | BQ278240.1 | - | 1 - 2 | 48 | 70 | |
| BU569345.1 | - | 1 - 2 | 60 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001317982.1 transcripts.
Set Membership: |
|

RPL26L1
Genomic Map