Assay Details
Gene Symbol: FAM57A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318008.1 | NP_001304937.1 | 2 - 3 | 307 | 71 | |
GenBank mRNA | CN259418.1 | - | 2 - 3 | 316 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
