Assay Details
| Gene Symbol: GAL3ST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318112.1 | NP_001305041.1 | 1 - 2 | 36 | 72 | |
| NM_001318114.1 | NP_001305043.1 | 1 - 2 | 36 | 72 | ||
| GenBank mRNA | BP276494.1 | - | 1 - 2 | 36 | 72 | |
| DA381099.1 | - | 1 - 2 | 33 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001318112.1, NM_001318114.1 transcripts.
Set Membership: |
|

GAL3ST1
Genomic Map