Assay Details
| Gene Symbol: SNX33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318146.1 | NP_001305075.1 | 1 - 2 | 1297 | 62 | |
| GenBank mRNA | AK091291.1 | - | 1 - 2 | 549 | 62 | |
More Information
Set Membership: |
|

SNX33
Genomic Map