Assay Details
| Gene Symbol: AQP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170.2 | NP_001161.1 | 4 - 5 | 605 | 107 | |
| NM_001318156.1 | NP_001305085.1 | 3 - 4 | 487 | 107 | ||
| NM_001318157.1 | NP_001305086.1 | 4 - 5 | 923 | 107 | ||
| NM_001318158.1 | NP_001305087.1 | 4 - 5 | 605 | 107 | ||
| XM_005251453.3 | XP_005251510.1 | 5 - 6 | 705 | 107 | ||
| XM_006716765.3 | XP_006716828.1 | 3 - 4 | 366 | 107 | ||
| XM_011517866.2 | XP_011516168.1 | 4 - 5 | 515 | 107 | ||
| XM_011517867.2 | XP_011516169.1 | 2 - 3 | 145 | 107 | ||
| XM_017014700.1 | XP_016870189.1 | 4 - 5 | 806 | 107 | ||
| XM_017014706.1 | XP_016870195.1 | 5 - 6 | 636 | 107 | ||
| GenBank mRNA | AB006190.1 | - | 4 - 5 | 442 | 107 | |
| AK297885.1 | - | 4 - 5 | 739 | 107 | ||
| BC062701.1 | - | 3 - 4 | 314 | 30324263 | 107 | |
More Information
Set Membership: |
|

AQP7
Genomic Map