Assay Details
Gene Symbol: LUZP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318840.1 | NP_001305769.1 | 1 - 2 | 129 | 95 | |
NM_016383.4 | NP_057467.1 | 1 - 2 | 129 | 95 | ||
GenBank mRNA | AF124430.1 | - | 1 - 2 | 99 | 95 | |
AK093369.1 | - | 1 - 2 | 120 | 95 | ||
AK314765.1 | - | 1 - 2 | 118 | 95 | ||
BC128134.1 | - | 1 - 2 | 91 | 40112435 | 95 | |
BC128135.1 | - | 1 - 2 | 91 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK093369.1, NM_001318840.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
