Assay Details
| Gene Symbol: COX7A2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319040.1 | NP_001305969.1 | 2 - 3 | 284 | 103 | |
| GenBank mRNA | CD700827.1 | - | 2 - 3 | 269 | 103 | |
More Information
Set Membership: |
|

COX7A2L
Genomic Map