Assay Details
| Gene Symbol: NLRP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319967.1 | NP_001306896.1 | 1 | 426 | 110 | |
| GenBank mRNA | AK026387.1 | - | 1 | 72 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NLRP2B| Gene Symbol: NLRP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319967.1 | NP_001306896.1 | 1 | 426 | 110 | |
| GenBank mRNA | AK026387.1 | - | 1 | 72 | 110 | |
Set Membership: |
|