Assay Details
| Gene Symbol: RDH16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320108.1 | NP_001307037.1 | 1 - 2 | 2226 | 89 | |
| GenBank mRNA | AB209332.1 | - | 1 - 2 | 2226 | 89 | |
More Information
Set Membership: |
|

RDH16
Genomic Map