Assay Details
| Gene Symbol: WDR20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320130.1 | NP_001307059.1 | 2 - 3 | 509 | 109 | |
| XM_011537356.2 | XP_011535658.1 | 2 - 3 | 537 | 109 | ||
| GenBank mRNA | BM707311.1 | - | 2 - 3 | 413 | 109 | |
| DR423616.1 | - | 2 - 3 | 431 | 109 | ||
More Information
Set Membership: |
|

WDR20
Genomic Map