Assay Details
| Gene Symbol: ATP6V1G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320218.1 | NP_001307147.1 | 1 - 2 | 74 | 75 | |
| NM_133262.2 | NP_573569.1 | 1 - 2 | 74 | 75 | ||
| NM_133326.1 | NP_579872.1 | 1 - 2 | 74 | 75 | ||
| GenBank mRNA | BF509031.1 | - | 4 - 5 | 618 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320218.1, NM_133262.2, NM_133326.1 transcripts.
Set Membership: |
|

ATP6V1G3
Genomic Map