Assay Details
| Gene Symbol: ATP6V1G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320218.1 | NP_001307147.1 | 2 - 3 | 193 | 117 | |
| XM_011509186.2 | XP_011507488.1 | 1 - 2 | 205 | 117 | ||
| GenBank mRNA | BC101129.2 | - | 1 - 2 | 111 | 40014378 | 117 |
More Information
Set Membership: |
|

ATP6V1G3
Genomic Map