Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320586.1 | NP_001307515.1 | 3 - 4 | 409 | 73 | |
| XM_017005406.1 | XP_016860895.1 | 4 - 5 | 755 | 73 | ||
| XM_017005407.1 | XP_016860896.1 | 3 - 4 | 674 | 73 | ||
| XM_017005411.1 | XP_016860900.1 | 4 - 5 | 755 | 73 | ||
| XM_017005412.1 | XP_016860901.1 | 4 - 5 | 755 | 73 | ||
| XM_017005413.1 | XP_016860902.1 | 4 - 5 | 755 | 73 | ||
| GenBank mRNA | BU543943.1 | - | 3 - 4 | 409 | 73 | |
More Information
Set Membership: |
|

ACYP2
Genomic Map