Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320588.1 | NP_001307517.1 | 2 - 3 | 255 | 91 | |
| GenBank mRNA | BF671963.1 | - | 2 - 3 | 254 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320588.1 transcripts.
Set Membership: |
|

ACYP2
Genomic Map