Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320611.1 | NP_001307540.1 | 6 - 7 | 1283 | 92 | |
| NM_002266.3 | NP_002257.1 | 6 - 7 | 925 | 92 | ||
| GenBank mRNA | AK024833.1 | - | 6 - 7 | 793 | 92 | |
| AK058111.1 | - | 6 - 7 | 794 | 92 | ||
| AK291954.1 | - | 6 - 7 | 792 | 92 | ||
| BC005978.1 | - | 6 - 7 | 792 | 4105058 | 92 | |
| BC013257.1 | - | 6 - 7 | 788 | 92 | ||
| BC034443.1 | - | 6 - 7 | 751 | 92 | ||
| BC053343.1 | - | 6 - 7 | 4450470 | 92 | ||
| BC067848.1 | - | 6 - 7 | 1024 | 4825235 | 92 | |
| BC146905.1 | - | 6 - 7 | 779 | 9056833 | 92 | |
| BT006665.1 | - | 5 - 6 | 663 | 92 | ||
| U09559.1 | - | 6 - 7 | 795 | 92 | ||
| U28386.1 | - | 6 - 7 | 795 | 92 | ||
More Information
Set Membership: |
|

KPNA2
Genomic Map