Assay Details
| Gene Symbol: LYPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321234.1 | NP_001308163.1 | 2 - 3 | 359 | 92 | |
| GenBank mRNA | AK130005.1 | - | 2 - 3 | 198 | 92 | |
More Information
Set Membership: |
|

LYPD1
Genomic Map