Assay Details
| Gene Symbol: DEGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321542.1 | NP_001308471.1 | 2 - 3 | 1063 | 90 | |
| NM_003676.3 | NP_003667.1 | 2 - 3 | 953 | 90 | ||
| XM_017002648.1 | XP_016858137.1 | 2 - 3 | 993 | 90 | ||
| GenBank mRNA | AF002668.1 | - | 2 - 3 | 895 | 90 | |
| AF466375.1 | - | 2 - 3 | 904 | 90 | ||
| AK308232.1 | - | 2 - 3 | 920 | 90 | ||
| AY423730.1 | - | 2 - 3 | 901 | 90 | ||
| BC000961.2 | - | 2 - 3 | 935 | 3450936 | 90 | |
More Information
Set Membership: |
|

DEGS1
Genomic Map