Assay Details
| Gene Symbol: NUTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322038.1 | NP_001308967.1 | 2 - 3 | 342 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001322038.1 transcripts.
Set Membership: |
|

NUTF2
Genomic Map