Assay Details
| Gene Symbol: NUTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322041.1 | NP_001308970.1 | 1 - 2 | 459 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001322041.1 transcripts.
Set Membership: |
|

NUTF2
Genomic Map