Assay Details
| Gene Symbol: ADRA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322502.1 | NP_001309431.1 | 1 - 2 | 1572 | 80 | |
| GenBank mRNA | AY491778.1 | - | 1 - 2 | 1322 | 80 | |
More Information
Set Membership: |
|

ADRA1A
Genomic Map