Assay Details
| Gene Symbol: NRSN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323679.1 | NP_001310608.1 | 5 - 6 | 900 | 63 | |
| GenBank mRNA | BP251173.1 | - | 5 - 6 | 521 | 63 | |
More Information
Set Membership: |
|

NRSN2
Genomic Map