Assay Details
| Gene Symbol: CTNNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323987.1 | NP_001310916.1 | 1 - 2 | 200 | 115 | |
| NM_001324004.1 | NP_001310933.1 | 1 - 2 | 200 | 115 | ||
| NM_001324008.1 | NP_001310937.1 | 1 - 2 | 200 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001323987.1, NM_001324004.1, NM_001324008.1 transcripts.
Set Membership: |
|

CTNNA1
Genomic Map