Assay Details
| Gene Symbol: SERPINB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276490.1 | NP_001263419.1 | 5 - 6 | 696 | 88 | |
| NM_002640.3 | NP_002631.3 | 6 - 7 | 874 | 88 | ||
| NM_198833.1 | NP_942130.1 | 6 - 7 | 891 | 88 | ||
| XM_017025792.1 | XP_016881281.1 | 4 - 5 | 498 | 88 | ||
| GenBank mRNA | AK300391.1 | - | 5 - 6 | 644 | 88 | |
| AK313782.1 | - | 6 - 7 | 791 | 88 | ||
| BX571754.1 | - | 6 - 7 | 782 | 88 | ||
| L40377.1 | - | 6 - 7 | 805 | 88 | ||
More Information
Set Membership: |
|

SERPINB8
Genomic Map