Assay Details
| Gene Symbol: B3GALNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003781.3 | NP_003772.1 | 4 - 5 | 412 | 72 | |
| NM_033169.2 | NP_149359.1 | 4 - 5 | 385 | 72 | ||
| XM_005247859.4 | XP_005247916.1 | 5 - 6 | 475 | 72 | ||
| XM_005247861.3 | XP_005247918.1 | 6 - 7 | 595 | 72 | ||
| XM_005247862.3 | XP_005247919.1 | 5 - 6 | 453 | 72 | ||
| XM_017007405.1 | XP_016862894.1 | 4 - 5 | 378 | 72 | ||
| XM_017007409.1 | XP_016862898.1 | 4 - 5 | 517 | 72 | ||
| XM_017007411.1 | XP_016862900.1 | 4 - 5 | 526 | 72 | ||
| GenBank mRNA | AB050856.1 | - | 4 - 5 | 328 | 72 | |
| AF132731.1 | - | 1 - 2 | 41 | 72 | ||
| AF154848.1 | - | 4 - 5 | 361 | 72 | ||
| AK022491.1 | - | 4 - 5 | 364 | 72 | ||
| AY359049.1 | - | 3 - 4 | 287 | 72 | ||
| BC047618.1 | - | 4 - 5 | 307 | 5259171 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB050856.1, AF132731.1, AF154848.1, AK022491.1, AY359049.1, BC047618.1, NM_003781.3, NM_033169.2, XM_005247859.4, XM_005247861.3, XM_005247862.3, XM_017007405.1, XM_017007409.1 transcripts.
Set Membership: |
|

B3GALNT1
Genomic Map