Assay Details
| Gene Symbol: XYLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005108.3 | NP_005099.2 | 1 - 2 | 169 | 93 | |
| XM_011534325.2 | XP_011532627.1 | 1 - 2 | 150 | 93 | ||
| XM_011534327.1 | XP_011532629.1 | 1 - 2 | 150 | 93 | ||
| XM_011534328.2 | XP_011532630.1 | 1 - 2 | 150 | 93 | ||
| XM_011534329.1 | XP_011532631.1 | 1 - 2 | 150 | 93 | ||
| XM_011534330.2 | XP_011532632.1 | 1 - 2 | 150 | 93 | ||
| XM_017007595.1 | XP_016863084.1 | 1 - 2 | 69 | 93 | ||
| XM_017007598.1 | XP_016863087.1 | 1 - 2 | 150 | 93 | ||
| XM_017007599.1 | XP_016863088.1 | 1 - 2 | 150 | 93 | ||
| XM_017007600.1 | XP_016863089.1 | 1 - 2 | 155 | 93 | ||
| GenBank mRNA | AB015046.1 | - | 1 - 2 | 180 | 93 | |
| AK025728.1 | - | 1 - 2 | 121 | 93 | ||
| AK293325.1 | - | 1 - 2 | 74 | 93 | ||
| AK314386.1 | - | 1 - 2 | 117 | 93 | ||
| BC030231.2 | - | 1 - 2 | 138 | 93 | ||
| BC137076.1 | - | 1 - 2 | 101 | 9021073 | 93 | |
| BC137080.1 | - | 1 - 2 | 101 | 9021077 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK293325.1, XM_017007595.1, XM_017007600.1 transcripts.
Set Membership: |
|

XYLB
Genomic Map